Product Details
- SNP ID
-
rs138817702
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11770965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAAATTGAATGCAATCTCT[A/G]GAAAGCAAAGATATATAACCAATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF441
PubMed Links
Gene Details
- Gene
- ZNF441
- Gene Name
- zinc finger protein 441
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152355.2 |
|
Intron |
|
|
NP_689568.2 |
View Full Product Details