Product Details

SNP ID
rs138870150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48663536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACATATTGCAGTAGTTTTGACA[C/G]TGAGGGTCTGGGGAGGGTCAGGCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NTN5 PubMed Links

Gene Details

Gene
NTN5
Gene Name
netrin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145807.2 1240 Silent Mutation CAC,CAG H344Q NP_665806.1
XM_011526443.2 1240 Missense Mutation CAC,CAG H344Q XP_011524745.1
XM_017026274.1 1240 Missense Mutation CAC,CAG H189Q XP_016881763.1
Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details