Product Details

SNP ID
rs139445983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57634694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGAATGGGATCTCCTTGATGA[A/G]GCTCAGAAACACCTGTACTTCGATG
Phenotype
MIM: 604076 MIM: 601856
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
There are no transcripts associated with this gene.

Gene
ZNF211
Gene Name
zinc finger protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265597.2 375 Silent Mutation GAA,GAG E65E NP_001252526.1
NM_001265598.2 375 Silent Mutation GAA,GAG E52E NP_001252527.1
NM_001265599.2 375 UTR 5 NP_001252528.1
NM_001265600.2 375 UTR 5 NP_001252529.1
NM_001322306.1 375 Intron NP_001309235.1
NM_006385.4 375 Silent Mutation GAA,GAG E65E NP_006376.2
NM_198855.3 375 Silent Mutation GAA,GAG E52E NP_942152.1

View Full Product Details