Product Details

SNP ID
rs139859732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43626167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCGTCTGCTTGCTGTGTCCGGA[A/G]GGCAGCGCCTGGTTCTGCGCCTCAC
Phenotype
MIM: 609744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145296.1 579 Silent Mutation CCC,CCT P207P NP_660339.1
XM_005258620.2 579 Silent Mutation CCC,CCT P207P XP_005258677.1
XM_017026452.1 579 Silent Mutation CCC,CCT P188P XP_016881941.1
Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

View Full Product Details