Product Details
- SNP ID
-
rs140511415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18782926 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCACCCTGGTCCCTAGGCTTGCC[A/G]CAGCTGATGGGTCTCATAGTCCTCT
- Phenotype
-
MIM: 600310
MIM: 607536
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COMP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61752496] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COMP
- Gene Name
- cartilage oligomeric matrix protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000095.2 |
2299 |
Missense Mutation |
CGG,TGG |
R755W |
NP_000086.2 |
- Gene
- CRTC1
- Gene Name
- CREB regulated transcription coactivator 1
View Full Product Details