Product Details

SNP ID
rs140519538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19202498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCAGCCCCGGCGACTGGAGAA[A/G]CCACCCTGAAACTGGATCTGCAGCT
Phenotype
MIM: 616601 MIM: 600826 MIM: 608719 MIM: 603200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BORCS8 PubMed Links

Gene Details

Gene
BORCS8
Gene Name
BLOC-1 related complex subunit 8
There are no transcripts associated with this gene.

Gene
BORCS8-MEF2B
Gene Name
BORCS8-MEF2B readthrough
There are no transcripts associated with this gene.

Gene
NCAN
Gene Name
neurocan
There are no transcripts associated with this gene.

Gene
NR2C2AP
Gene Name
nuclear receptor 2C2 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300945.1 600 Silent Mutation GGC,GGT G69G NP_001287874.1
NM_176880.5 600 Silent Mutation GGC,GGT G69G NP_795361.1
XM_011527688.2 600 Silent Mutation GGC,GGT G69G XP_011525990.1
Gene
RFXANK
Gene Name
regulatory factor X associated ankyrin containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278727.1 600 Intron NP_001265656.1
NM_001278728.1 600 Intron NP_001265657.1
NM_003721.3 600 Intron NP_003712.1
NM_134440.2 600 Intron NP_604389.1
XM_005260134.4 600 Intron XP_005260191.1
XM_005260135.3 600 Intron XP_005260192.1
XM_005260136.4 600 Intron XP_005260193.1
XM_005260137.4 600 Intron XP_005260194.1
XM_006722930.3 600 Intron XP_006722993.1
XM_017027415.1 600 Intron XP_016882904.1
XM_017027416.1 600 Intron XP_016882905.1

View Full Product Details