Product Details

SNP ID
rs140631734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13099649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGGCAGCAGCTTCCTCAGCTC[C/G]GCGAAGGCGCCGTTAACGTTCTGCT
Phenotype
MIM: 151440 MIM: 164005 MIM: 611669
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYL1 PubMed Links

Gene Details

Gene
LYL1
Gene Name
LYL1, basic helix-loop-helix family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005583.4 1720 Silent Mutation GCC,GCG A171A NP_005574.2
XM_005259912.3 1720 Silent Mutation GCC,GCG A97A XP_005259969.1
XM_017026816.1 1720 Silent Mutation GCC,GCG A242A XP_016882305.1
XM_017026817.1 1720 Silent Mutation GCC,GCG A193A XP_016882306.1
Gene
NFIX
Gene Name
nuclear factor I X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271043.2 1720 Intron NP_001257972.1
NM_001271044.2 1720 Intron NP_001257973.1
NM_002501.3 1720 Intron NP_002492.2
XM_005259917.4 1720 Intron XP_005259974.1
XM_005259918.4 1720 Intron XP_005259975.1
XM_005259919.3 1720 Intron XP_005259976.1
XM_005259920.3 1720 Intron XP_005259977.1
XM_006722760.2 1720 Intron XP_006722823.1
XM_011528040.1 1720 Intron XP_011526342.1
XM_017026837.1 1720 Intron XP_016882326.1
Gene
TRMT1
Gene Name
tRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details