Product Details

SNP ID
rs141764075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48472383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACACACCCGAGGAGATCGCCC[A/G]CTTCCTGTACAAGGGCGAGGGGCTG
Phenotype
MIM: 602488 MIM: 603953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYTH2 PubMed Links

Gene Details

Gene
CYTH2
Gene Name
cytohesin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004228.6 593 Missense Mutation CAC,CGC H98R NP_004219.3
NM_017457.5 593 Missense Mutation CAC,CGC H98R NP_059431.1
XM_006723472.2 593 Missense Mutation CAC,CGC H120R XP_006723535.1
XM_006723473.2 593 Missense Mutation CAC,CGC H120R XP_006723536.1
Gene
KCNJ14
Gene Name
potassium voltage-gated channel subfamily J member 14
There are no transcripts associated with this gene.

Gene
LOC105372430
Gene Name
uncharacterized LOC105372430
There are no transcripts associated with this gene.

View Full Product Details