Product Details
- SNP ID
-
rs141947949
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49583592 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGAGCTTCCTGAGTAGCCATACC[C/T]GGATTCCAAGAGCCAACCACTGGGA
- Phenotype
-
MIM: 616759
MIM: 616633
MIM: 604429
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NOSIP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs543865817] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NOSIP
- Gene Name
- nitric oxide synthase interacting protein
There are no transcripts associated with this gene.
- Gene
- PRR12
- Gene Name
- proline rich 12
There are no transcripts associated with this gene.
- Gene
- PRRG2
- Gene Name
- proline rich and Gla domain 2
View Full Product Details