Product Details

SNP ID
rs142773664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51058614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGATCTTTCCTGGGTAGACTTGA[C/T]GACACTCCTCATCTGAGCGAAGTTG
Phenotype
MIM: 605505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK13 PubMed Links

Gene Details

Gene
KLK13
Gene Name
kallikrein related peptidase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015596.1 612 Missense Mutation CAT,CGT H190R NP_056411.1

View Full Product Details