Product Details

SNP ID
rs142957625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9566834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACTGATCAACAAAGGCTTCCTC[A/G]CAGTCACTGTATTCAAAGGATTTGT
Phenotype
MIM: 194628
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF121 PubMed Links

Gene Details

Gene
ZNF121
Gene Name
zinc finger protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008727.3 511 Silent Mutation TGC,TGT C93C NP_001008727.1
NM_001308269.1 511 Silent Mutation TGC,TGT C93C NP_001295198.1
XM_017027239.1 511 Silent Mutation TGC,TGT C93C XP_016882728.1

View Full Product Details