Product Details

SNP ID
rs143679494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4816676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGAGGTAGGCTGAGTAGGCTG[C/T]GTTCAGTGCCGCCGCCTGCATCCGC
Phenotype
MIM: 607601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TICAM1 PubMed Links

Gene Details

Gene
TICAM1
Gene Name
toll like receptor adaptor molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182919.3 1970 Missense Mutation NP_891549.1

View Full Product Details