Product Details

SNP ID
rs143816968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16639004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGCTTTATTTCCCCTAACATTT[C/T]CACAGTTCCCTTGGCACAGTCCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMIM7 PubMed Links

Gene Details

Gene
SMIM7
Gene Name
small integral membrane protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300925.1 Intron NP_001287854.1
NM_024104.3 Intron NP_077009.2

View Full Product Details