Product Details
- SNP ID
-
rs143824265
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4658021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCTGGTCACAGCTCAGTGCGC[A/G]ATGCCTTGGCCACAATCACCAGCTT
- Phenotype
-
MIM: 606746
MIM: 615869
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MYDGF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12683] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYDGF
- Gene Name
- myeloid derived growth factor
- Gene
- TNFAIP8L1
- Gene Name
- TNF alpha induced protein 8 like 1
There are no transcripts associated with this gene.
View Full Product Details