Product Details
- SNP ID
-
rs143947857
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51367906 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACCGCCGAGGAAGAGGAAGGCG[C/T]TCGTGGTCTGGCCCCGCAGCGACTC
- Phenotype
-
MIM: 130410
MIM: 606008
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDND2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74538792] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLDND2
- Gene Name
- claudin domain containing 2
- Gene
- ETFB
- Gene Name
- electron transfer flavoprotein beta subunit
There are no transcripts associated with this gene.
- Gene
- NKG7
- Gene Name
- natural killer cell granule protein 7
There are no transcripts associated with this gene.
View Full Product Details