Product Details

SNP ID
rs144269922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37151081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAATACTGAATGAGGGTTGGAT[A/G]TGACCAACTGTGGTAGATTCGAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF585A PubMed Links

Gene Details

Gene
ZNF585A
Gene Name
zinc finger protein 585A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288800.1 3077 UTR 3 NP_001275729.1
NM_152655.3 3077 UTR 3 NP_689868.1
NM_199126.2 3077 UTR 3 NP_954577.1

View Full Product Details