Product Details

SNP ID
rs144943269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49744294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCATGGAGAGGTCAGGCCCT[C/T]TTCGAGAGTTCTGAAATTGCCGCCC
Phenotype
MIM: 608253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSKS PubMed Links

Gene Details

Gene
TSKS
Gene Name
testis specific serine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021733.1 1309 Missense Mutation AAA,AGA K433R NP_068379.1
XM_011527201.1 1309 Missense Mutation AAA,AGA K433R XP_011525503.1
XM_011527202.1 1309 Missense Mutation AAA,AGA K433R XP_011525504.1
XM_011527203.2 1309 Missense Mutation AAA,AGA K433R XP_011525505.1

View Full Product Details