Product Details
- SNP ID
-
rs145129604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:50422871 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCACCTGTCCCAGCCACCCCCT[A/G]TGAAGCCTTCGACCCGGCAGCAGCC
- Phenotype
-
MIM: 174761
MIM: 606802
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POLD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61729811] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POLD1
- Gene Name
- DAN polymerase delta 1, catalytic subunit
There are no transcripts associated with this gene.
- Gene
- SPIB
- Gene Name
- Spi-B transcription factor
View Full Product Details