Product Details

SNP ID
rs145172873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2166783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCTGGTTGCAGGGTACAGTTCT[A/G]TACGTTTTGACAAACTCATTGGGTG
Phenotype
MIM: 607375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOT1L PubMed Links

Gene Details

Gene
DOT1L
Gene Name
DOT1 like histone lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032482.2 Intron NP_115871.1
XM_005259659.3 Intron XP_005259716.1
XM_005259660.3 Intron XP_005259717.1
XM_011528359.2 Intron XP_011526661.1
XM_011528360.1 Intron XP_011526662.1
XM_011528361.2 Intron XP_011526663.1
XM_017027366.1 Intron XP_016882855.1

View Full Product Details