Product Details

SNP ID
hCV165424681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54108053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGGGGCCAGCTCTGGAGTCAG[C/T]GCATTTCCTGCTCGGCGTCCATCCC
Phenotype
MIM: 603832 MIM: 606862 MIM: 606419 MIM: 609519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
There are no transcripts associated with this gene.

Gene
OSCAR
Gene Name
osteoclast associated, immunoglobulin-like receptor
There are no transcripts associated with this gene.

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
There are no transcripts associated with this gene.

Gene
TFPT
Gene Name
TCF3 fusion partner
There are no transcripts associated with this gene.

View Full Product Details