Product Details
- SNP ID
-
rs145817468
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8697844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGCTCCTTGCCCAGGGTGACCGT[A/G]CGCCCATCGGGCAGCTTCAGAGTCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTL9
PubMed Links
Gene Details
- Gene
- ACTL9
- Gene Name
- actin like 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178525.4 |
987 |
Silent Mutation |
CGC,CGT |
R286R |
NP_848620.3 |
View Full Product Details