Product Details

SNP ID
rs145884302
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37635462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATAGCCACGACTGAAAGTCTTC[A/C]CACATTCCTTACAATCATAAGGCTT
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ZFP30 PubMed Links

Gene Details

Gene
ZFP30
Gene Name
ZFP30 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320666.1 1401 Missense Mutation GGG,GTG G360V NP_001307595.1
NM_001320667.1 1401 Missense Mutation GGG,GTG G360V NP_001307596.1
NM_001320668.1 1401 Missense Mutation GGG,GTG G360V NP_001307597.1
NM_001320669.1 1401 Missense Mutation GGG,GTG G360V NP_001307598.1
NM_001320670.1 1401 Missense Mutation GGG,GTG G359V NP_001307599.1
NM_001320671.1 1401 Missense Mutation GGG,GTG G359V NP_001307600.1
NM_014898.2 1401 Missense Mutation GGG,GTG G360V NP_055713.1

View Full Product Details