Product Details

SNP ID
rs145899020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12939147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCACTTAGACGGGTGGACTTC[C/T]CGCTGGATCGAATCCAAACACAAGT
Phenotype
MIM: 109091 MIM: 602918 MIM: 600061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALR PubMed Links

Gene Details

Gene
CALR
Gene Name
calreticulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004343.3 185 Silent Mutation TCC,TCT S35S NP_004334.1
Gene
FARSA
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
There are no transcripts associated with this gene.

Gene
MIR6515
Gene Name
microRNA 6515
There are no transcripts associated with this gene.

Gene
RAD23A
Gene Name
RAD23 homolog A, nucleotide excision repair protein
There are no transcripts associated with this gene.

View Full Product Details