Product Details

SNP ID
rs145953497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35803025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGAAGCCACCATGAGGTGGCAC[A/G]TGGGGCCATGGCGGGCCACGTGCGT
Phenotype
MIM: 616377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01529 PubMed Links

Gene Details

Gene
LINC01529
Gene Name
long intergenic non-protein coding RNA 1529
There are no transcripts associated with this gene.

Gene
PRODH2
Gene Name
proline dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021232.1 1158 Missense Mutation ACG,ATG T428M NP_067055.1
XM_017027084.1 1158 Missense Mutation ACG,ATG T352M XP_016882573.1
XM_017027085.1 1158 Missense Mutation ACG,ATG T352M XP_016882574.1

View Full Product Details