Product Details

SNP ID
rs145953617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35538320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTGGCCAGATTTGGACGCATC[A/G]GTCGCCTGGTCCTGCGCGCCTGCAT
Phenotype
MIM: 609169 MIM: 609969 MIM: 613585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAPDHS PubMed Links

Gene Details

Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014364.4 271 Missense Mutation AGT,GGT S87G NP_055179.1
XM_017026579.1 271 Missense Mutation AGT,GGT S86G XP_016882068.1
Gene
SBSN
Gene Name
suprabasin
There are no transcripts associated with this gene.

Gene
TMEM147
Gene Name
transmembrane protein 147
There are no transcripts associated with this gene.

Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details