Product Details

SNP ID
rs146090389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37413515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCACACTCATTACATTCATA[A/G]GGTTTTTCACCTGTATGAATTCTAA
Phenotype
MIM: 613904
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF569 PubMed Links

Gene Details

Gene
ZNF569
Gene Name
zinc finger protein 569
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152484.2 1284 Silent Mutation CCC,CCT P381P NP_689697.2
XM_006723046.2 1284 Silent Mutation CCC,CCT P405P XP_006723109.1
XM_006723047.3 1284 Silent Mutation CCC,CCT P405P XP_006723110.1
XM_006723048.3 1284 Silent Mutation CCC,CCT P405P XP_006723111.1
XM_011526538.2 1284 Silent Mutation CCC,CCT P405P XP_011524840.1
XM_011526539.2 1284 Silent Mutation CCC,CCT P381P XP_011524841.1
XM_017026376.1 1284 Silent Mutation CCC,CCT P381P XP_016881865.1
XM_017026377.1 1284 Silent Mutation CCC,CCT P381P XP_016881866.1
XM_017026378.1 1284 Silent Mutation CCC,CCT P345P XP_016881867.1
XM_017026379.1 1284 Silent Mutation CCC,CCT P222P XP_016881868.1
XM_017026380.1 1284 Silent Mutation CCC,CCT P222P XP_016881869.1
XM_017026381.1 1284 Silent Mutation CCC,CCT P222P XP_016881870.1
XM_017026382.1 1284 Silent Mutation CCC,CCT P222P XP_016881871.1

View Full Product Details