Product Details
- SNP ID
-
rs146198329
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49478972 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCGCCTCCTGCAGGAGACCTCC[A/G]AGCAGCTGGTGGCGCTGAAGCCCTG
- Phenotype
-
MIM: 613358
MIM: 600007
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH16A1
PubMed Links
Gene Details
- Gene
- ALDH16A1
- Gene Name
- aldehyde dehydrogenase 16 family member A1
There are no transcripts associated with this gene.
- Gene
- FLT3LG
- Gene Name
- fms related tyrosine kinase 3 ligand
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001204502.1 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
NP_001191431.1 |
NM_001204503.1 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
NP_001191432.1 |
NM_001278637.1 |
442 |
Missense Mutation |
AAG,GAG |
K54E |
NP_001265566.1 |
NM_001278638.1 |
442 |
Missense Mutation |
AAG,GAG |
K54E |
NP_001265567.1 |
NM_001459.3 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
NP_001450.2 |
XM_005258680.4 |
442 |
Missense Mutation |
AAG,GAG |
K118E |
XP_005258737.3 |
XM_005258681.4 |
442 |
Missense Mutation |
AAG,GAG |
K118E |
XP_005258738.3 |
XM_005258682.4 |
442 |
Intron |
|
|
XP_005258739.3 |
XM_005258683.4 |
442 |
Intron |
|
|
XP_005258740.3 |
XM_006723116.3 |
442 |
Intron |
|
|
XP_006723179.2 |
XM_011526675.2 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
XP_011524977.1 |
XM_011526676.2 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
XP_011524978.1 |
XM_011526677.2 |
442 |
Missense Mutation |
AAG,GAG |
K136E |
XP_011524979.1 |
XM_011526678.2 |
442 |
Missense Mutation |
AAG,GAG |
K118E |
XP_011524980.1 |
XM_011526680.2 |
442 |
Intron |
|
|
XP_011524982.1 |
XM_011526682.2 |
442 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011524984.1 |
XM_017026532.1 |
442 |
Intron |
|
|
XP_016882021.1 |
XM_017026533.1 |
442 |
Missense Mutation |
AAG,GAG |
K99E |
XP_016882022.1 |
XM_017026534.1 |
442 |
Missense Mutation |
AAG,GAG |
K99E |
XP_016882023.1 |
XM_017026535.1 |
442 |
Missense Mutation |
AAG,GAG |
K36E |
XP_016882024.1 |
- Gene
- RPL13A
- Gene Name
- ribosomal protein L13a
There are no transcripts associated with this gene.
View Full Product Details