Product Details

SNP ID
rs146615949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35903843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGTGGCTGCTGATGCGGTGGCA[C/T]CGCTGCTCATCGTGGGGGCGGTGTT
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007469.1 295 Missense Mutation CCG,TCG P61S NP_001007470.1
NM_014266.3 295 Missense Mutation CCG,TCG P61S NP_055081.1
XM_017026193.1 295 Silent Mutation CAC,CAT H101H XP_016881682.1
Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173514.1 295 Intron NP_001166985.1
NM_001173515.1 295 Intron NP_001166986.1
NM_003332.3 295 Intron NP_003323.1
NM_198125.2 295 Intron NP_937758.1

View Full Product Details