Product Details

SNP ID
rs146635380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42403022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCTGCTTCAGACACACTGCGG[C/T]GCATCGGCTCTGAGAGAGGGAGAGC
Phenotype
MIM: 151750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPE PubMed Links

Gene Details

Gene
LIPE
Gene Name
lipase E, hormone sensitive type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005357.3 1694 Missense Mutation CAC,CGC H851R NP_005348.2
XM_005258937.3 1694 Missense Mutation CAC,CGC H775R XP_005258994.1
XM_005258938.4 1694 Missense Mutation CAC,CGC H596R XP_005258995.1
XM_005258939.3 1694 Missense Mutation CAC,CGC H613R XP_005258996.2
XM_005258940.3 1694 Missense Mutation CAC,CGC H550R XP_005258997.1
XM_006723218.2 1694 Missense Mutation CAC,CGC H550R XP_006723281.1
XM_017026810.1 1694 Missense Mutation CAC,CGC H550R XP_016882299.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101930071
Gene Name
uncharacterized LOC101930071
There are no transcripts associated with this gene.

View Full Product Details