Product Details

SNP ID
rs146718961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTGACTGAAGCCCTTACAACA[C/G]ACATCACATATGTAAGGCCTCTCTC
Phenotype
MIM: 604749
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 6304 Silent Mutation GTC,GTG V715V NP_004225.3
XM_005259412.4 6304 Silent Mutation GTC,GTG V711V XP_005259469.1

View Full Product Details