Product Details

SNP ID
rs146736409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54355398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCATACGTCACCTCCTGGGAA[C/T]TCCCTGCAGCCAGGGCCTAAGAGGG
Phenotype
MIM: 602992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAIR1 PubMed Links

Gene Details

Gene
LAIR1
Gene Name
leukocyte associated immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289023.2 1044 Missense Mutation AAT,AGT N227S NP_001275952.2
NM_001289025.2 1044 Missense Mutation AAT,AGT N244S NP_001275954.2
NM_001289026.2 1044 Missense Mutation AAT,AGT N238S NP_001275955.2
NM_001289027.2 1044 Missense Mutation AAT,AGT N227S NP_001275956.2
NM_002287.5 1044 Missense Mutation AAT,AGT N245S NP_002278.2
NM_021706.4 1044 Missense Mutation AAT,AGT N228S NP_068352.2
XM_017026803.1 1044 Intron XP_016882292.1

View Full Product Details