Product Details

SNP ID
rs146867163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45163949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTGCCACGGAGGCGGTGACCA[C/T]GCTCTCAATGCCCAGGGTATAGAGG
Phenotype
MIM: 610396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKPD1 PubMed Links

Gene Details

Gene
NKPD1
Gene Name
NTPase, KAP family P-loop domain containing 1
There are no transcripts associated with this gene.

Gene
TRAPPC6A
Gene Name
trafficking protein particle complex 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270891.1 440 Missense Mutation ATG,GTG M139V NP_001257820.1
NM_001270892.1 440 UTR 3 NP_001257821.1
NM_001270893.1 440 UTR 3 NP_001257822.1
NM_024108.2 440 Missense Mutation ATG,GTG M153V NP_077013.1
XM_017027295.1 440 UTR 3 XP_016882784.1

View Full Product Details