Product Details
- SNP ID
-
hCV165453212
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41847825 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCACCTGATGAGGAGAGGGGAAG[C/G]CTCTCCCAAAGCTCCCAACCACTTC
- Phenotype
-
MIM: 614806
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DMRTC2
PubMed Links
Gene Details
- Gene
- DMRTC2
- Gene Name
- DMRT like family C2
There are no transcripts associated with this gene.
- Gene
- LYPD4
- Gene Name
- LY6/PLAUR domain containing 4
There are no transcripts associated with this gene.
View Full Product Details