Product Details

SNP ID
rs147383567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15728851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCCCAAAGGTCCCCACTCTCAG[C/G]AGGGTGACACCCTGATGGCCACCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10H2 PubMed Links

Gene Details

Gene
OR10H2
Gene Name
olfactory receptor family 10 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013939.2 828 Missense Mutation CAG,GAG Q270E NP_039227.1

View Full Product Details