Product Details
- SNP ID
-
rs147588657
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:42588859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGTGGTGTGGCAGGCATAGGATC[C/T]GCTGTTCTTTGTAGTGATGTTGGGG
- Phenotype
-
MIM: 615747
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEACAM8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35599593] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEACAM8
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 8
- Gene
- LIPE-AS1
- Gene Name
- LIPE antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details