Product Details

SNP ID
rs147753936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55898830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACACAGTAGCTTCACTCCATCA[A/T]CCTGAAGATCATTTTCTCCCAAATC
Phenotype
MIM: 609660
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NLRP13 PubMed Links

Gene Details

Gene
NLRP13
Gene Name
NLR family pyrin domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321057.1 2922 Missense Mutation GAT,GTT D966V NP_001307986.1
NM_176810.2 2922 Missense Mutation GAT,GTT D966V NP_789780.2

View Full Product Details