Product Details

SNP ID
rs147759603
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47611265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGATAATATTTAGAAGAAGCG[A/G]CCAGATTTTTTTTTTCCCTGGGAGC
Phenotype
MIM: 605690
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GLTSCR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs545176661] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GLTSCR1
Gene Name
glioma tumor suppressor candidate region gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015711.3 Intron NP_056526.3
XM_005258833.4 Intron XP_005258890.1
XM_006723180.3 Intron XP_006723243.1
XM_011526882.2 Intron XP_011525184.1
XM_011526883.2 Intron XP_011525185.1

View Full Product Details