Product Details

SNP ID
rs147860527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58556731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGCCCTCGAGGTCAATGTTGGG[A/G]TGATAGACCATTGTCTCACACTTCA
Phenotype
MIM: 610893 MIM: 194550 MIM: 601742 MIM: 603173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP2A PubMed Links

Gene Details

Gene
CHMP2A
Gene Name
charged multivesicular body protein 2A
There are no transcripts associated with this gene.

Gene
MIR6807
Gene Name
microRNA 6807
There are no transcripts associated with this gene.

Gene
MZF1
Gene Name
myeloid zinc finger 1
There are no transcripts associated with this gene.

Gene
MZF1-AS1
Gene Name
MZF1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TRIM28
Gene Name
tripartite motif containing 28
There are no transcripts associated with this gene.

Gene
UBE2M
Gene Name
ubiquitin conjugating enzyme E2 M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003969.3 898 Silent Mutation CAC,CAT H101H NP_003960.1

View Full Product Details