Product Details

SNP ID
rs148027395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8697954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGGTGCTTAATGTTCTCCACTA[A/G]GTCCAGGTCCTGCTGTCCCAGGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL9 PubMed Links

Gene Details

Gene
ACTL9
Gene Name
actin like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178525.4 877 Silent Mutation CTA,TTA L250L NP_848620.3

View Full Product Details