Product Details

SNP ID
rs148124007
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49739819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCCTGTTTTGGGGGGGTTCCT[A/G]CACTGCTGCCTCCCCCCATTGTTCC
Phenotype
MIM: 608253
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TSKS PubMed Links

Gene Details

Gene
TSKS
Gene Name
testis specific serine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021733.1 1747 Missense Mutation GCA,GTA A579V NP_068379.1
XM_011527201.1 1747 Missense Mutation GCA,GTA A604V XP_011525503.1
XM_011527202.1 1747 UTR 3 XP_011525504.1
XM_011527203.2 1747 UTR 3 XP_011525505.1

View Full Product Details