Product Details

SNP ID
rs148237208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49660789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACACAGAACCAGAGGGCATAG[C/T]GTGGTGAGCGTCCGCTTCCTTCCGT
Phenotype
MIM: 610837 MIM: 603734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL2L12 PubMed Links

Gene Details

Gene
BCL2L12
Gene Name
BCL2 like 12
There are no transcripts associated with this gene.

Gene
IRF3
Gene Name
interferon regulatory factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197122.1 1333 Silent Mutation ACA,ACG T346T NP_001184051.1
NM_001197123.1 1333 Missense Mutation CAC,CGC H306R NP_001184052.1
NM_001197124.1 1333 Missense Mutation CAC,CGC H214R NP_001184053.1
NM_001197125.1 1333 Missense Mutation CAC,CGC H195R NP_001184054.1
NM_001197126.1 1333 Missense Mutation CAC,CGC H195R NP_001184055.1
NM_001197127.1 1333 Missense Mutation CAC,CGC H68R NP_001184056.1
NM_001197128.1 1333 Missense Mutation CAC,CGC H68R NP_001184057.1
NM_001571.5 1333 Missense Mutation CAC,CGC H341R NP_001562.1
XM_006723197.1 1333 Silent Mutation ACA,ACG T346T XP_006723260.1
XM_006723198.1 1333 Silent Mutation ACA,ACG T346T XP_006723261.1
XM_006723200.1 1333 Silent Mutation ACA,ACG T311T XP_006723263.1
XM_006723201.1 1333 Silent Mutation ACA,ACG T200T XP_006723264.1
XM_006723202.2 1333 Silent Mutation ACA,ACG T200T XP_006723265.1
XM_017026766.1 1333 Missense Mutation CAC,CGC H341R XP_016882255.1
XM_017026767.1 1333 Missense Mutation CAC,CGC H341R XP_016882256.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details