Product Details

SNP ID
rs148260314
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36514848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACATGCATAGGGTTTGGTTCCTG[G/T]ATGATTTTTCTGGTGTCTGATGATG
Phenotype
MIM: 613749
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012756.2 1307 Missense Mutation ACA,CCA T131P NP_001012774.1
NM_001166036.1 1307 Missense Mutation ACA,CCA T131P NP_001159508.1
NM_001166037.1 1307 Missense Mutation ACA,CCA T131P NP_001159509.1
NM_001166038.1 1307 Missense Mutation ACA,CCA T131P NP_001159510.1
XM_011526897.2 1307 Missense Mutation ACA,CCA T131P XP_011525199.1
XM_017026741.1 1307 Missense Mutation ACA,CCA T131P XP_016882230.1

View Full Product Details