Product Details

SNP ID
rs148332097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19624497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTACTTTTCTACAGCCAGGACA[G/T]TGCAGGACTCACAGCCTAAACCATC
Phenotype
MIM: 609694 MIM: 605110 MIM: 608127
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GMIP PubMed Links

Gene Details

Gene
GMIP
Gene Name
GEM interacting protein
There are no transcripts associated with this gene.

Gene
LPAR2
Gene Name
lysophosphatidic acid receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004720.5 954 Missense Mutation AAT,ACT N272T NP_004711.2
XM_011528421.2 954 Missense Mutation AAT,ACT N272T XP_011526723.1
XM_017027468.1 954 Missense Mutation AAT,ACT N272T XP_016882957.1
XM_017027469.1 954 Missense Mutation AAT,ACT N272T XP_016882958.1
Gene
PBX4
Gene Name
PBX homeobox 4
There are no transcripts associated with this gene.

View Full Product Details