Product Details

SNP ID
rs148522375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12878428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAAGCTCTGGAAAACAGCCCC[A/G]GCCTGGGATGTGAGTGTGATGCTGC
Phenotype
MIM: 126350 MIM: 600599 MIM: 612256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNASE2 PubMed Links

Gene Details

Gene
DNASE2
Gene Name
deoxyribonuclease II, lysosomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001375.2 809 Silent Mutation GCC,GCT A221A NP_001366.1
Gene
KLF1
Gene Name
Kruppel like factor 1
There are no transcripts associated with this gene.

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
There are no transcripts associated with this gene.

View Full Product Details