Product Details

SNP ID
rs148804845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13774558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCAGCCTCTGAAAAGAATCG[A/G]GGCCCAAGAAAAGGCGGTAAGGAGC
Phenotype
MIM: 615105
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf53 PubMed Links

Gene Details

Gene
C19orf53
Gene Name
chromosome 19 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014047.2 116 Silent Mutation CGA,CGG R27R NP_054766.1
Gene
MRI1
Gene Name
methylthioribose-1-phosphate isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528355.2 116 Intron XP_011526657.1
XM_011528356.2 116 Intron XP_011526658.1
XM_011528357.2 116 Intron XP_011526659.1

View Full Product Details