Product Details

SNP ID
rs148923526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29528442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTTGCATTTTAGGTAACAAAT[A/G]AGGATGCAACTAAAATCAGCGTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC284395 PubMed Links

Gene Details

Gene
LOC284395
Gene Name
uncharacterized LOC284395
There are no transcripts associated with this gene.

Gene
VSTM2B
Gene Name
V-set and transmembrane domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146339.1 897 Missense Mutation AAG,GAG K93E NP_001139811.1
XM_005258856.3 897 Intron XP_005258913.1
XM_011526902.2 897 Missense Mutation AAG,GAG K93E XP_011525204.1
XM_011526903.2 897 Intron XP_011525205.1
XM_011526904.2 897 Missense Mutation AAG,GAG K47E XP_011525206.1
XM_011526905.2 897 Intron XP_011525207.1

View Full Product Details