Product Details
- SNP ID
-
rs148968963
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37669497 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACCAGTATGAATTCTTTGATGT[C/T]GAGTAAGATGTGCACTTTGCCTAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF781
PubMed Links
Gene Details
- Gene
- ZNF781
- Gene Name
- zinc finger protein 781
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152605.3 |
1394 |
Missense Mutation |
AAC,GAC |
N218D |
NP_689818.2 |
View Full Product Details