Product Details
- SNP ID
-
rs149294852
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14840950 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTTCTTGAAAAATAGCCTTTCT[A/G]TTGCTTTCCTCTGAAGAATGTTTTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR7A10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3752193,rs8100008] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR7A10
- Gene Name
- olfactory receptor family 7 subfamily A member 10
There are no transcripts associated with this gene.
- Gene
- OR7A5
- Gene Name
- olfactory receptor family 7 subfamily A member 5
There are no transcripts associated with this gene.
- Gene
- OR7C1
- Gene Name
- olfactory receptor family 7 subfamily C member 1
There are no transcripts associated with this gene.
View Full Product Details