Product Details

SNP ID
rs149691289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37634288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGCTTAAATTCAGGTTCAATA[C/T]TTTTGGCAAGATGTGTATATCTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP30 PubMed Links

Gene Details

Gene
ZFP30
Gene Name
ZFP30 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320666.1 2575 UTR 3 NP_001307595.1
NM_001320667.1 2575 UTR 3 NP_001307596.1
NM_001320668.1 2575 UTR 3 NP_001307597.1
NM_001320669.1 2575 UTR 3 NP_001307598.1
NM_001320670.1 2575 UTR 3 NP_001307599.1
NM_001320671.1 2575 UTR 3 NP_001307600.1
NM_014898.2 2575 UTR 3 NP_055713.1

View Full Product Details