Product Details

SNP ID
rs149754029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54311632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATGAACCTGTCGAATCTCAGC[A/C]GTGAGCCACACTGGAGGGTCACGTT
Phenotype
MIM: 606047
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LILRA5 PubMed Links

Gene Details

Gene
LILRA5
Gene Name
leukocyte immunoglobulin like receptor A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021250.3 578 Intron NP_067073.1
NM_181879.2 578 Missense Mutation CAG,CCG Q165P NP_870994.1
NM_181985.3 578 Intron NP_871714.1
NM_181986.2 578 Missense Mutation CAG,CCG Q153P NP_871715.1

View Full Product Details